Imprimer
Phusion® High-Fidelity PCR kit
Marques : FINNZYMES REAGENTS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F-553L F-553S
F-553LEA 312 EUR
F-553L F-553S
Phusion® High-Fidelity PCR kit
Réactifs pour acides nucléiques End-point PCR Enzymes and Kits
Phusion® High-Fidelity PCR kit offers extreme performance for all major PCR applications. Incorporating Phusion® DNA polymerase, the Phusion® High-Fidelity PCR kit contains all the reagents necessary, including lambda DNA control template and primers for 1,3 kb and 10 kb amplicons. The template amount is sufficient for performing 20 control reactions in 50 μl volume or 50 control reactions in 20 μl volume.
Informations de livraison: Each kit contains Phusion® DNA polymerase (2 U/µl), 5X Phusion® HF Buffer, 5X Phusion® GC Buffer, dNTP mix (10 mM/each), MgCl₂ solution (50 mM), control lambda template (0,5 ng/µl), 1,3 kb primers (4 µM each), 10 kb primers (4 µM each), and DNA size standard.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Application
Learn more