Imprimer
Phusion® Flash high-fidelity PCR master mix
Marques : FINNZYMES REAGENTS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F-548L F-548S
F-548LEA 393 EUR
F-548L F-548S
Phusion® Flash high-fidelity PCR master mix
Réactifs pour acides nucléiques End-point PCR Enzymes and Kits
Phusion® Flash High-Fidelity PCR Master Mix is a 2X master mix enabling the use of extremely short PCR protocols without compromising either the fidelity or the yield in the reaction. The master mix utilises Phusion® Flash II DNA Polymerase, a proofreading DNA polymerase modified from Phusion® Hot Start II High-Fidelity DNA Polymerase. The fidelity of the Phusion® Flash II DNA Polymerase is 25-fold greater than Taq.