Imprimer
Phusion® High-Fidelity PCR Master Mix with HF Buffer
Marques : FINNZYMES REAGENTS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F-531L F-531S
F-531LEA 836 EUR
F-531L F-531S
Phusion® High-Fidelity PCR Master Mix with HF Buffer
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Phusion® High-Fidelity PCR Master Mix with HF Buffer is a convenient 2X mix containing Phusion® DNA Polymerase, nucleotides and optimised HF buffer including MgCl₂. Only template and primers need to be added by the user.
Informations de livraison: Each kit contains Phusion® DNA Polymerase, 2X Phusion® HF Buffer and 400 µM of each dNTP. The master mix supplies 1,5 mM MgCl₂ and 200 µM dNTP in final reaction concentration. A separate tube of DMSO is provided with the product.