Imprimer
Phusion® Hot Start II High-Fidelity DNA polymerase
Marques : FINNZYMES REAGENTS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F-549L F-549S F-549XL
F-549LEA 678 EUR
F-549L F-549S F-549XL
Phusion® Hot Start II High-Fidelity DNA polymerase
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Phusion® Hot Start High-Fidelity DNA polymerase allows the use of a wide range of primers, including those with low melting temperatures. Phusion® Hot Start High-Fidelity DNA polymerase is ideal for PCR applications from routine PCR to highly demanding applications, such as cloning and high throughput PCR. It incorporates a unique dsDNA-binding domain making this Pyrococcus-like proofreading DNA polymerase extremely robust, rapid, and accurate.
  • No annealing temperature limitations
  • Allows reaction set up at room temperature
  • Zero-time reactivation due to unique Hot Start technology
  • Fast PCR enzyme with extreme fidelity (over 6 times more accurate than Pfu)
  • Robust enzyme with high tolerance against PCR inhibitors requires minimal optimisation