Imprimer
Scrapers, SteriWare®
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8785H-110S 8785H-110 8785H-75 8785H-75S
SAMP8785H-110SEA 432 EUR
SAMP8785H-110S SAMP8785H-110 SAMP8785H-75 SAMP8785H-75S
Scrapers, SteriWare®
Raclettes
These disposable scrapers are made from high quality HDPE. They are ideal in the manufacture of creams, ointments and pastes.
  • Moulded & packed in a cleanroom
  • Made from FDA and EU conforming HDPE
  • BSE/TSE-free
  • Available as gamma irradiated sterile (SAL 10-6)
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling
Full batch traceability
Cdt: Individually bagged.