Imprimer
Economy scoop
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A304-450 A304-1000 A304-120 A304-1500 A304-2000
SAMPA304-450EA 78.9 EUR
SAMPA304-450 300-0660 300-0661 300-0662 300-0663
Economy scoop
Scoops
These scoops made from 304 stainless steel are ideal for general industrial use.
  • High polish
  • Light duty construction
  • Round handle
Please note: This scoop is not suitable for GMP applications.