Imprimer
Distributeurs adaptables sur flacons, LABmax airless HF
Fournisseur: Witeg
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
WITG5.370.825EA 1870 EUR
WITG5.370.825 WITG5.370.824 WITG5.370.823
Distributeurs adaptables sur flacons, LABmax airless HF
Distributeurs pour flacons
LABmax Airless HF is a special dispenser for hydrofluoric acid (HF). The components which have direct contact with the acid are made of HF-resistant materials. LABmax Airless HF is equipped with an automatic air-purging system. All dispensers are ready to use after no more than two dosings.
  • High quality precision instrument
  • Highest accuracy and precision, with error ranges better than DIN EN ISO 8655-5 requirements
  • 100% drip-free due to a unique anti-drip system
  • 360° rotating valve block for added safety
  • One-handed operation
  • Flexibility to use different bottle adapters
  • Easy to clean, completely autoclavable at 121 °C
  • Individual certificate and serial number
  • DE-M marked according to the German calibration law
The robust and stable dispenser is made of high quality materials, including platinum spring, ruby valve balls, sapphire valve, PTFE.
Certifications: ISO 9001, CE certified.
Informations de livraison: Supplied with LABmax airless HF, suction tube, discharge tube, instruction manual, individual certificate, serial number, thread adapter (10,0 ml: A25, A28, A45, S40 | 25,0/50,0 ml: A25, A28, A32/45, S40).