Imprimer
Distributeurs adaptables sur flacons, LABmax airless
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5.370.001 5.370.801 5.370.802 5.370.803 5.370.804 5.370.805 5.370.002 5.370.003 5.370.004 5.370.005 5.370.006
613-9111EA 357 EUR
613-9111 WITG5.370.801 WITG5.370.802 WITG5.370.803 WITG5.370.804 WITG5.370.805 WITG5.370.002 WITG5.370.003 WITG5.370.004 WITG5.370.005 WITG5.370.006
Distributeurs adaptables sur flacons, LABmax airless
Distributeurs pour flacons
Dispensers have a unique direct displacement piston system that is chemical-resistant and provides safe, smooth dispensing.
  • Unique closed-circuit air purging system
  • Reagent guard valve prevents leakage
  • Exclusive 360° swivel turn
  • Quick, accurate volume adjustment
  • Unique piston design
Units are designed with a unique air purging system that prevents loss of reagents and eliminates dripping or splashing of media. An exclusive reagent guard valve prohibits accidental discharge and eliminates the need for capping the discharge tube when the dispenser is not in use. The volume adjustment knob allows quick and easy setting, and locks the volume into place. The discharge tube can be turned on a 360° swivel, allowing convenient dispensing and safe positioning over the reagent label. Basic dispensers have a PTFE system for broad general use. Choose a PTFE-lined dispenser for applications using hydrofluoric acid or a glass-lined dispenser for use with organic solvents. Dispensers disassemble easily for cleaning. Automatic calibration eliminates readjustment. Fully autoclavable to 121 °C.
Certifications: ISO 9001, CE certified.
Informations de livraison: Supplied with operation mode selector for air puring, filling and discharge tubes, thread adapters and instruction manual.