Imprimer
Mixing Measuring cylinders, tall form, class B, borosilicate glass
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3.750.100 3.750.010 3.750.003 3.750.005
612-4967EA 119 EUR
612-4967 WITG3.750.010 WITG3.750.003 WITG3.750.005
Mixing Measuring cylinders, tall form, class B, borosilicate glass
Éprouvettes Éprouvettes graduées
Borosilicate glass, class B, with PE stopper.
  • Calibrated to contain 'ln'
  • Hexagonal plastic base, NS ground socket
  • Brown graduations
Certifications: DIN EN ISO 4788