Imprimer
Distillation apparatus, micro
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2.996.000 2.996.005
WITG2.996.000EA 373 EUR
WITG2.996.000 WITG2.996.005
Distillation apparatus, micro
Composants de distillation Pièces pour distillation
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Short path distilling apparatus, micro, complete.