Imprimer
Lames bistouris pour greffe de peau
Fournisseur: SWANN MORTON
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9935 9940
SWAN9935EA 114 EUR
SWAN9935 SWAN9940
Lames bistouris pour greffe de peau
Bistouris et pièces détachées Lames de bistouri
Stainless steel. The 158 mm skin graft blades are used in the surgical removal of a patch of healthy skin from one area of the patient's body, usually the buttocks or inner thigh, which is then transplanted to another.
  • Sterile
  • Fit the Braithwaite (9901), Cobbett (9902) and Watson (9903) Skin Graft handles
Skin grafts may be recommended for burns, specific surgeries that may require skin grafts for healing to occur and for areas of prior infection with extensive skin loss.