Imprimer
Syringes for Shimadzu GC autosamplers
Fournisseur: HAMILTON BONADUZ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
202630 202640 202643 93898-01
549-1113EA 248 EUR
549-1113 549-1114 549-1058 549-1060
Syringes for Shimadzu GC autosamplers
Seringues Seringues de chromatographie
These syringes are used for GC autosamplers.
  • Removable needle
  • Cemented needle termination with metal flange and glass flange