Imprimer
Powder funnels, PP
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8023P-100 8023P-150 8023P-250 8023P-350
SAMP8023P-100EA 4.4 EUR
SAMP8023P-100 221-0317 SAMP8023P-250 SAMP8023P-350
Powder funnels, PP
Entonnoirs Entonnoirs à poudre
Reusable powder funnel, made of PP, crevice free, with wide diameter discharge spout to enable powder to be discharged without catching.
  • Good chemical resistance
  • Good temperature resistance
  • Ideal for loading powder into drums, containers or kegs