Imprimer
Funnels with short stem
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8022P-100 8022P-200
SAMP8022P-100EA 13.8 EUR
SAMP8022P-100 SAMP8022P-200
Funnels with short stem
Entonnoirs Entonnoirs pour la filtration Entonnoirs filtrants
These liquid funnels have PP construction. Crevice free.
  • Good chemical resistance
  • Good temperature resistance
  • Easy to clean