Imprimer
Closure with brush for bottles
Fournisseur: RIXIUS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8-1090-18-57 8-1090-18-77
RIXI8-1090-18-57EA 52.8 EUR
RIXI8-1090-18-57 RIXI8-1090-18-77
Closure with brush for bottles
Capuchons Bouchons pour flacons
Closures with brush made of HDPE.
  • Brush with goat black hair of about 57 mm length
  • For 10 ml round bottles