Imprimer
Mixing measuring cylinders, tall form, class A, borosilicate glass, VOLAC
Fournisseur: Poulten & Graf
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
R264/B/2 R264/D/2 R264/E/2 R264/F/2 R264/G/2 R264/I/2 R264/J/S R264/WAC/B/2 R264/WAC/E/2 R264/WAC/F/2 R264/WAC/G/2 R264/WAC/H/2 R264/WAC/I/2 R264/WAC/J/S R264/H/2
POULR264/B/2EA 65.9 EUR
POULR264/B/2 POULR264/D/2 POULR264/E/2 POULR264/F/2 POULR264/G/2 POULR264/I/2 POULR264/J/S POULR264/WAC/B/2 POULR264/WAC/E/2 POULR264/WAC/F/2 POULR264/WAC/G/2 POULR264/WAC/H/2 POULR264/WAC/I/2 POULR264/WAC/J/S POULR264/H/2
Mixing measuring cylinders, tall form, class A, borosilicate glass, VOLAC
Éprouvettes Éprouvettes graduées
Borosilicate glass, class A.
  • White ceramic glazed ink scale
  • Hexagonal glass base, NS ground socket
  • Available without or with work certificate
Certifications: ISO4788 - BS 604 - DIN 12680