Imprimer
Échantillonneur composite Ex, pour liquides inflammables
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5072A-500 5072A-1000 5072B-500
SAMP5072A-500EA 2440 EUR
SAMP5072A-500 SAMP5072A-1000 SAMP5072B-500
Échantillonneur composite Ex, pour liquides inflammables
Échantillonneurs Préleveurs de liquides
Essential for flammable liquids as a heavy-duty sampler.
  • Metallic parts manufactured from stainless steel
  • Unique double action to sample from a set distance
  • Can be plunged until the assembly touches the bottom of the container
Specially designed for the environment throughout the world for both chemical and environmental sampling.
Certifications: Conforms to FDA CFR 177.2600.