Imprimer
Separating funnels, conical
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2.676.001 2.676.002 2.676.500
WITG2.676.001EA 305 EUR
WITG2.676.001 WITG2.676.002 WITG2.676.500
Separating funnels, conical
Entonnoirs Ampoules à décanter
Separating funnels, conical, made of DURAN® borosilicate glass 3.3, graduated, with Witaflo-needle valve stopcock and PTFE-needle spindle.
  • Easy to read graduation
  • With plastic stopper
  • Very good chemical and temperature resistance