Imprimer
Separating funnels, Squibb
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2.647.001 2.647.050 2.647.100 2.647.250 2.647.500
WITG2.647.001EA 169 EUR
WITG2.647.001 WITG2.647.050 WITG2.647.100 WITG2.647.250 WITG2.647.500
Separating funnels, Squibb
Entonnoirs Ampoules à décanter
Separating funnels, Squibb, made of borosilicate glass 3.3, with NS stopcock plugs and solid lass key. With PE-stopper.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance