Imprimer
Cleaning brushes for samplers
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8020A-1100 8020A-1600 8020N-1100 8020N-1600 8020A-2100 8020A-600 8020J-1100 8020J-1600 8020J-2100 8020J-600 8020N-2100 8020N-600
300-0492EA 135 EUR
300-0492 300-0493 300-0513 300-0514 300-0494 300-0495 300-0496 300-0497 300-0498 300-0499 300-0515 300-0516
Cleaning brushes for samplers
Brushes
Brushes with nylon bristles in a twisted stainless steel wire frame and aluminium handle. Ideal for cleaning both stainless steel and plastic samplers.
  • Available in a range of lengths and diameters