Imprimer
Nessler cylinders
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2.150.050 2.150.150 2.151.100 2.150.100 2.151.050
WITG2.150.050EA 34.1 EUR
WITG2.150.050 WITG2.150.150 WITG2.151.100 WITG2.150.100 WITG2.151.050
Nessler cylinders
Tubes Tubes de Nessler
Colour comparison cylinders according to Nessler for comparators, with optically plane bottom.