Imprimer
Bottles, narrow or wide neck, square or rectangular, with thread
Fournisseur: RIXIUS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2-E0-0500-60 2-E0-0250-60 2-M0-2000-60 2-M0-1000-80 2-M0-2000-80 2-V0-0100-40 2-S0-0050-18 2-D0-0100-25 2-E0-1000-25 2-U0-1000-60 2-M0-1500-80 2-H0-0500-40 2-E0-1000-40 2-OP0-1000-60 2-V0-0125-40 2-D0-0030-25 2-UO-250-80 2-E0-1000-60 2-D0-0050-25-K 2-E0-1000-25WA 2-V0-0250-40I 2-V0-0250-25WA 2-H0-0500-25FO 2-M0-2500-80 2-U0-1000-80
RIXI2-E0-0500-60EA 1.8 EUR
RIXI2-E0-0500-60 RIXI2-E0-0250-60 RIXI2-M0-2000-60 RIXI2-M0-1000-80 RIXI2-M0-2000-80 RIXI2-V0-0100-40 RIXI2-S0-0050-18 RIXI2-D0-0100-25 RIXI2-E0-1000-25 RIXI2-U0-1000-60 RIXI2-M0-1500-80 RIXI2-H0-0500-40 RIXI2-E0-1000-40 RIXI2-OP0-1000-60 RIXI2-V0-0125-40 RIXI2-D0-0030-25 RIXI2-UO-250-80 RIXI2-E0-1000-60 RIXI2-D0-0050-25-K RIXI2-E0-1000-25WA RIXI2-V0-0250-40I RIXI2-V0-0250-25WA RIXI2-H0-0500-25FO RIXI2-M0-2500-80 RIXI2-U0-1000-80
Bottles, narrow or wide neck, square or rectangular, with thread
Flacons Flacons en plastique
HDPE, natural.
  • Rigid and shatter-proof
  • Resistant to water, aqueous solutions and most chemicals
  • Excellent water vapour resistance
  • Ribbed sides for secure grip
Square or rectangular bottles with large filling opening for packaging and storage of liquids, powders and granulates.
Informations de commande: Screw caps must be ordered seperately.