Imprimer
Dropping funnels, cylindrical, with pressure balance
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.895.001 1.895.022 1.895.023 1.895.002 1.895.021
WITG1.895.001EA 107 EUR
WITG1.895.001 WITG1.895.022 WITG1.895.023 WITG1.895.002 WITG1.895.021
Dropping funnels, cylindrical, with pressure balance
Entonnoirs Ampoules de coulée
Dropping funnels, cylindrical, made of borosilicate glass, stopcock with glass key. With drip nozzle.
  • Graduated
  • Very good chemical resistance
  • Very good temperature resistance
With screw-thread retaling nut.