Imprimer
Dropping funnels, cylindrical, with pressure balance
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.869.025 1.869.050 1.869.100 1.869.250
WITG1.869.025EA 253 EUR
WITG1.869.025 WITG1.869.050 WITG1.869.100 WITG1.869.250
Dropping funnels, cylindrical, with pressure balance
Entonnoirs Ampoules de coulée
Dropping funnels, cylindrical, made of borosilicate glass, with needle valve stopcock with PTFE-needle valve.
  • Graduated
  • With NS cone and socket
  • Very good chemical and temperature resistance
With screw-thread retaining nut.