Imprimer
Dropping funnels, cylindrical, without pressure balance
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.812.050 1.812.100
WITG1.812.050EA 97.6 EUR
WITG1.812.050 WITG1.812.100
Dropping funnels, cylindrical, without pressure balance
Entonnoirs Ampoules de coulée
Dropping funnels, cylindrical, made of borosilicate glass 3.3, with plug with PTFE key, NS cone and socket.
  • Easy to read graduations
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes