Imprimer
Dropping funnels, cylindrical, without pressure balance
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.802.050 1.802.100 1.806.250 1.806.500
WITG1.802.050EA 84.9 EUR
WITG1.802.050 WITG1.802.100 WITG1.806.250 WITG1.806.500
Dropping funnels, cylindrical, without pressure balance
Entonnoirs Ampoules de coulée
Cylindrical dropping funnels, made of borosilicate glass with screw-thread retaining nut.
  • With glass key stopcock and graduation
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Certifications: DIN 12567