Imprimer
Immersion coolers
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.786.219 1.786.214 1.786.229.S
WITG1.786.219EA 70.2 EUR
WITG1.786.219 WITG1.786.214 WITG1.786.229.S
Immersion coolers
Condenseurs
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For immersion into distillation assemblies etc. Can be used to regulate refluxing by adjusting the flow of air or water through it. Water connection with two glass olives.