Imprimer
Condensers, Air
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.780.050 1.782.050
WITG1.780.050EA 23 EUR
WITG1.780.050 WITG1.782.050
Condensers, Air
Condenseurs
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With standard ground joint cone.