Imprimer
Condensers, Allihn, for Soxleth extractors
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2.200.034 2.200.045 2.200.060 2.200.029
WITG2.200.034EA 130 EUR
WITG2.200.034 WITG2.200.045 WITG2.200.060 WITG2.200.029
Condensers, Allihn, for Soxleth extractors
Condenseurs
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With NS cone. Water connection with glass nozzles.