Imprimer
Condensers, Allihn
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.720.250 1.720.400 1.723.250 1.724.250 1.727.400 1.724.400 1.728.400
WITG1.720.250EA 96.2 EUR
WITG1.720.250 WITG1.720.400 WITG1.723.250 WITG1.724.250 WITG1.727.400 WITG1.724.400 WITG1.728.400
Condensers, Allihn
Condenseurs
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With or without ground joints. Water connection with glass nozzles.