Imprimer
Flasks, Erlenmeyer, with standard ground joint
Fournisseur: GLASWARENFABRIK KARL HECHT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
41915006 41915014 41915012 41915020 41915018 41915024 41915030 41915034 41915036 41915042 41915046 41915048 41915052 41915056 41918006 41918012 41918020 41918018 41918024 41918030 41918036 41918042 41918048
HECH41915006EA 40.3 EUR
HECH41915006 HECH41915014 HECH41915012 HECH41915020 HECH41915018 HECH41915024 HECH41915030 HECH41915034 HECH41915036 HECH41915042 HECH41915046 HECH41915048 HECH41915052 HECH41915056 HECH41918006 HECH41918012 HECH41918020 HECH41918018 HECH41918024 HECH41918030 HECH41918036 HECH41918042 HECH41918048
Flasks, Erlenmeyer, with standard ground joint
Ballons Flacons Erlenmeyers
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Volume graduations and labelling area.