Imprimer
Glove liners, RU 711
Fournisseur: HONEYWELL SÉCURITÉ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
RU711/6 RU711/8
DALLRU711/6EA 32.88 EUR
DALLRU711/6 DALLRU711/8
Glove liners, RU 711
Gants
These gloves made from 93% cotton and 7% elasthane are ideal for precision work and finishing operations in mechanical environments, clock-making, jewellery, photography. Also suitable for handlings requiring a clean glove without any plush.
  • Cotton interlock ensures the glove's mechanical performance while absorbing hand perspiration for added comfort
  • Glove is ambidextrous and thus more cost effective
  • Tight knit makes gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • Ribbed at the wrist to ensure a good level of wrist comfort, thus eliminating risks of getting it caught
  • White colour enhances the glove's hygiene and cleanliness properties
Certifications: Cat. I