Imprimer
Screws with cylindrical head, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H1128-26 H1128-14 H1128-22 H1128-18
BOHLH1128-26EA 28.9 EUR
BOHLH1128-26 BOHLH1128-14 BOHLH1128-22 BOHLH1128-18
Screws with cylindrical head, BOLA
Fixations Accouplements
PTFE, white.
  • Temperature resistance: from -200 °C to +250 °C
  • Universal chemical resistance
  • FDA conform
  • Similar to DIN 84/DIN EN ISO 1207