Imprimer
N-Phényl-3-(phosphonooxy)naphtalène-2-carboxamide ≥98% (par HPLC), grade Technique, Sigma-Aldrich®
Marques : SIGMA ALDRICH MICROSCOPY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): 7-Bromo-3-hydroxy-2-naphthoic-o-anisidide phosphate, Naphthol AS-BI Phosphoric acid solution, Naphthol AS-BI-Phosphate, 6-Bromo-3-((2-methoxyphenyl)carbamoyl)naphthalen-2-yl dihydrogen phosphate
M-Clarity™ (Niveau de Qualité) = MQ200
N2125-100MG N2125-1G N2125-500MG N2125-5G
N2125-100MGEA 110 EUR
N2125-100MG N2125-1G N2125-500MG N2125-5G
N-Phényl-3-(phosphonooxy)naphtalène-2-carboxamide ≥98% (par HPLC), grade Technique, Sigma-Aldrich®
N-Phényl-3-(phosphonooxy)naphtalène-2-carboxamide
Substrate for the histochemical demonstration of acid and alkaline phosphatase. Substrate for the fluorometric assay of acid and alkaline phosphatases. Demonstration of acid phosphatase in paraffin embedded tissue. Cytofluorometric assay of acid phosphatase in single living cells.
  • Suitable for haematology
  • Suitable for histology
Formule: C₁₈H₁₅BrNO₆P
Poids moléculaire: 452.1926 g/mol
Température de stockage: Congélateur
Numéro MDL: MFCD00004058
Numéro CAS: 1919-91-1
EINECS: 217-645-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96cafb60079b0bd0a25c6e289e787950690ecd94&vid=69276bc3177bccaa86ee3353c2578faea6bb9e1d&prid=web&clid=SAPDAM