Imprimer

Creatine phosphate disodium salt, Millipore®

Marques : Merck Millipore Calbiochem
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Synonyme(s): Phosphocreatine disodium hydrate, Sodium 2-(1-methyl-3-phosphonatoguanidino)acetate hydrate(2:1:x), N-(Imino[phosphonoamino]methyl)-N-methylglycine hydrate, Phosphocreatine disodium salt hydrate

2380-25 2380-5
2380-25EA 437 EUR
2380-25 2380-5
Creatine phosphate disodium salt, Millipore®
Phosphocreatine sel de sodium hydraté
Formule: C₄H₈N₃Na₂O₅P·H₂O
Numéro MDL: MFCD00150191
Numéro CAS: 19333-65-4
EINECS: 213-074-6

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR