Imprimer
Tissue pulveriser, Bessman type
Fournisseur: Spectrum Laboratories
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
189475 189476 189470
SPEC189475EA 2150 EUR
SPEC189475 SPEC189476 SPEC189470
Tissue pulveriser, Bessman type
Homogénéisateurs Broyeurs pour tissus
The stainless steel Bessman tissue pulveriser consists of a two-component mortar with handles and a pestle, specifically designed for pulverising 10 - 1000 mg of tissue.
After chilling the pulveriser in liquid nitrogen, the included 28 cm lead hammer is used on the mortar to completely homogenise the inserted sample. The sample can then easily be transferred without touching or thawing by removing the mortar and inverting the pestle assembly over a test tube.
Informations de livraison: The small and medium pulveriser includes 10 test tubes and the large pulveriser includes one culture jar.