Imprimer
Échantillonneurs, U-D Master
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1230A-1200 1230B-600 1230B-1200 1230B-1500 1230B-900 1230A-1500 1230A-1800 1230A-300 1230A-600 1230A-900 1230B-1800 1230C-1200 1230C-1500 1230C-1800 1230C-900
SAMP1230A-1200EA 8760 EUR
SAMP1230A-1200 SAMP1230B-600 SAMP1230B-1200 SAMP1230B-1500 SAMP1230B-900 SAMP1230A-1500 SAMP1230A-1800 SAMP1230A-300 SAMP1230A-600 SAMP1230A-900 SAMP1230B-1800 SAMP1230C-1200 SAMP1230C-1500 SAMP1230C-1800 SAMP1230C-900
Échantillonneurs, U-D Master
Échantillonneurs Préleveurs de solides
U-D Master is designed for sampling free flowing powders. By simply changing the sample insert cells, different volumes can be sampled.
  • Operation of the unit dose powder sampler is very easy
  • Push the sampler: At the required depth rotate the handle, which will take the sample
  • After removing the sampler the sample inserts can be removed for easy emptying of the sample
Single version takes one sample from each location.
Double version takes two samples from each location.
Triple version takes three samples from each location.