Imprimer
ECM mimetic cultureware plates, Corning® PureCoat™
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
356270 356271
734-2494EA 374 EUR
734-2494 734-2495
ECM mimetic cultureware plates, Corning® PureCoat™
Microplaques Microplaques pour culture cellulaire
Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor and primary cell types.
  • Available peptides: Human collagen I and human fibronectin
  • Delivers similar adhesion capabilities as the native protein, whilst being completely synthetic and animal-free
  • Ideal for researchers looking for defined (SF/XF/AF) culture conditions of cells in basic and applied clinical research, vaccine production and biomanufacturing
Corning PureCoat cultureware is coated with biologically active, synthetic, animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent, cost-effective alternative to self-coated extracellular peptides. The proprietary covalent linkage orients the peptides for optimal binding and signaling.