Imprimer
Adapters, extension
Fournisseur: COWIE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PC713.025050GF PC713.050050GF PC713.050075GF PC713.075075GF PC713.075100GF PC713.100100GF PC713.100150GF
COWIPC713.025050GFEA 35.9 EUR
COWIPC713.025050GF COWIPC713.050050GF COWIPC713.050075GF COWIPC713.075075GF COWIPC713.075100GF COWIPC713.100100GF COWIPC713.100150GF
Adapters, extension
Raccords et connecteurs
NPT extension adapters are manufactured in glass filled PTFE for improved mechanical strength.
  • Chemically inert
  • Reduce contamination and simplify cleaning
  • Non-scratch
Male/female NPT extension adapters optimise the use of NPT entry ports by the provision of additional port sizes and raising the height of the entry port from the lid. This gives even more flexibility of assembly and enables a greater range of fittings to be used - this is especially the case for 60mm lids with multiple ports where the use of an extension adapter allows the use of a central stirrer guide.