Imprimer
Automatic burette with glass key, Class AS, Pellet type, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
243182705 243182754 243183307 243183356 243183607 243183656
612-5213EA 318 EUR
612-5213 612-5214 612-5215 612-5216 612-5217 612-5218
Automatic burette with glass key, Class AS, Pellet type, DURAN®
Burettes
Burette, automatic, Pellet, DURAN® borosilicate glass 3.3, clear, class AS.
  • With Schellbach stripe and main graduations as circular divisions
  • With glass key
  • Side-positioned standard ground stopcock
  • 30 seconds waiting time
With printed retrace code (batch identification).
Certifications: DIN EN ISO 385
Informations de livraison: Supplied with reservoir bottle and rubber air pump