Imprimer
Cartouches d'extraction, haute pureté, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2814-199 2814-432 2814-300 2814-259 2814-533
WHAT2814-199EA 453 EUR
WHAT2814-199 515-9012 WHAT2814-300 WHAT2814-259 WHAT2814-533
Cartouches d'extraction, haute pureté, Whatman™
Extracteurs Cartouches d'extraction
Cytiva's Whatman 100% borosilicate glass microfiber (HP-GF) thimbles are designed to fit most Soxhlet extraction units. They are well-suited to pollution monitoring when analysis of hot and acidic gases is required.
  • Resistant to temperatures up to 500 °C
  • Binder-free, no chemical additives
  • Designed to fit most commercially available Soxhlet extractors
  • Effective when used with solvents incompatible with cellulose materials
Glass Soxhlet extraction thimble applications:
Analysis of hot and acidic gases during emission control and smoke stack gas monitoring
Analysis of pesticide residues using Soxhlet extraction
Determination of oil/fat content of foods
Analysis of oil and grease in solid wastes
Whatman brand high-purity glass thimbles are available in a range of sizes and wall thicknesses for optimal use in a variety of air sampling and Soxhlet extraction units.