Imprimer
Drums, combination
Fournisseur: DUPERTHAL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5.10.006 5.10.010
DUEH5.10.006EA 51.3 EUR
DUEH5.10.006 DUEH5.10.010
Drums, combination
Barils, tonneaux, fûts
These combination drum has a outer jacket steel sheet with blue enamel and inner bubble polyethylene.
  • Tight closure
  • BAM allowed
  • Transport approval for all packaging groups
Container identification according to GefStoffV and BGV A8.