Imprimer
Portoirs de refroidissement
Fournisseur: GLW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
G01 G01S G02 G02S
GLWAG01EA 21.6 EUR
GLWAG01 GLWAG01S GLWAG02 GLWAG02S
Portoirs de refroidissement
Portoirs Portoirs de refroidissement
These cooling racks are made of polypropylene for test tubes that require continued cooling.
  • Sturdy construction
  • Autoclavable up to +121 °C
  • Cover must be ordered seperately
Remove the insert, add chrushed ice, reinsert and start working. Use in freezer down to −90° C