Imprimer
Transport trolleys for glassware, MetroMax i®
Fournisseur: Metro International
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
GWBX3-1 GWBX3-2 GWTX3-2 GWTX3-1
139-0062EA 1490 EUR
139-0062 139-0063 139-0070 139-0069
Transport trolleys for glassware, MetroMax i®
Trolleys
MetroMax i® glassware carts are available in two and four basket capacities and are ideal for transporting glassware safely in the laboratory.
  • The solid bottom shelf provides spill retention and can be removed from shelf frame for easy cleaning
Informations de commande: Castors must be ordered separately.