Imprimer
PIG® Original absorbent socks
Fournisseur: NEW PIG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
404 204 PIG205 PIG209 PIG218 PIG219 PIG220
NEWP404EA 138 EUR
NEWP404 NEWP204 NEWPPIG205 NEWPPIG209 NEWPPIG218 NEWPPIG219 NEWPPIG220
PIG® Original absorbent socks
Matériaux absorbants
Formable socks hug corners and surround machine bases to absorb machine leaks, contain puddles and help keep workers safe.
  • Tough and economical; good choice for everyday leak protection
  • Contained corncob filler pulls liquids from the floor for maximum absorbency whilst making cleanup quick and easy
  • Polypropylene skin resists tearing; reduces dust and holds in liquid, even when saturated
  • Absorbs and retains oils, coolants, solvents and water
  • Corncob filler is light-weight and economical to handle general, everyday tasks
Can be incinerated after use to reduce waste or for fuels blending. 99% pre-consumer recycled cellulose filler. With Clip-&-Fit Absorbent Sock you can create your own custom lengths of PIG® Original Sock - pull exact length you need out of dispenser box, "clip" ends with included cable ties (100 included), cut the material and "fit" your new sock perfectly into place.