Imprimer
PIG® BLUE absorbent socks
Fournisseur: NEW PIG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4048 PIG201 PIG202 PIG203 2048 MA1097 PIG217
121-0500EA 184 EUR
121-0500 NEWPPIG201 NEWPPIG202 NEWPPIG203 NEWP2048 NEWPMA1097 NEWPPIG217
PIG® BLUE absorbent socks
Matériaux absorbants
Surround spills to prevent spreading. Extra-fine vermiculite filler pulls in liquid and hugs floors to stay in place.
  • Heaviest sock, features extra-dense construction to hug floors and stay in place whilst containing spills
  • Fine-grade vermiculite filler quickly absorbs leaks, drips and spills; great for spill response or absorbing machine leaks
  • Stitch-bonded, polypropylene skin resists bursting; reduces dust and holds in liquid, even when saturated
  • Absorbs and retains oils, coolants, solvents and water
Ideal for diking and damming liquids. Can be air-dried for reuse when soaked with water.