Imprimer
Dissecting trays
Fournisseur: NICKEL ELECTRO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
625 626
NICK625EA 116 EUR
NICK625 NICK626
Dissecting trays
Plateaux/Récipients/Pots Plateaux à dissection
Dissecting trays made of aluminium.
  • Good chemical resistance
  • Good temperature resistance
  • Easy to clean