Imprimer
Dissecting trays
Fournisseur: NICKEL ELECTRO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
695 697 698
NICK695EA 166 EUR
NICK695 NICK697 NICK698
Dissecting trays
Plateaux/Récipients/Pots Plateaux à dissection
Stainless steel.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy handling
For use in laboratories and schools.