Imprimer
Crucibles, stainless steel
Fournisseur: NICKEL ELECTRO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
123 124 126
NICK123EA 28.9 EUR
NICK123 NICK124 NICK126
Crucibles, stainless steel
Creusets
These stainless steel 304 crucibles with bright finish are much harder and last longer than porcelain types.
  • Non-magnetic
  • Shatterproof
Informations de commande: Lids need to be ordered separately.