Imprimer
Capillary columns, Elite-5 for autosystem
Fournisseur: PerkinElmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PERCN9316274EA 1360 EUR
PERCN9316274 PERCN9316076 PERCN9316110 PERCN9316077 PERCN9316275 PERCN9316111 PERCN9316276 PERCN9316277 PERCN9316112 PERCN9316079 PERCN9316279 PERCN9316073 PERCN9316074 PERCN9316272 PERCN9316273 PERCN9316075 PERCN9316087 PERCN9316286 PERCN9316088 PERCN9316287 PERCN9316089 PERCN9316289 PERCN9316080 PERCN9316082 PERCN9316083 PERCN9316084 PERCN9316282 PERCN9316085 PERCN9316283 PERCN9316086 PERCN9316284 PERCN9316098 PERCN9316296 PERCN9316297 PERCN9316099 PERCN9316298 PERCN9316299 PERCN9316090 PERCN9316091 PERCN9316290 PERCN9316092 PERCN9316291 PERCN9316292 PERCN9316293 PERCN9316294 PERCN9316295 PERCN9316066 PERCN9316067 PERCN9316100 PERCN9316101 PERCN9316068 PERCN9316069 PERCN9316102 PERCN9316300 PERCN9316103 PERCN9316301 PERCN9316302 PERCN9316303 PERCN9316108
Capillary columns, Elite-5 for autosystem
Colonnes pour chromatographie
Elite-5 and Elite-5ht columns are suitable for semivolatiles, phenols, amines, residual solvents, drugs of abuse, pesticides, PCB congeners (e.g. Aroclor mixes) and solvent impurities.
Elite-5MS capillary columns are general-purpose columns – ideal for GCMS analysis of semivolatiles, polycyclic aromatic compounds, chlorinated hydrocarbons, phthalates, phenols, amines, organochlorine pesticides, organophosphorus pesticides, drugs, solvent impurities, and hydrocarbons.